Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500909.1 | XP_003500957.1 | 3 - 4 | 725 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Probe spans exons |