Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502655.2 | XP_003502703.1 | 6 - 7 | 1278 | 92 | |
XM_007643294.1 | XP_007641484.1 | 6 - 7 | 1083 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |