Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504987.2 | XP_003505035.1 | 3 - 4 | 295 | 61 | |
GenBank mRNA | JP056546.1 | - | 517 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |