Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504987.2 | XP_003505035.1 | 7 - 8 | 629 | 97 | |
GenBank mRNA | JP056546.1 | - | 851 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |