Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644011.1 | XP_007642201.1 | 1 - 2 | 56 | 58 | |
XM_007644012.1 | XP_007642202.1 | 1 - 2 | 122 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |