Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514760.2 | XP_003514808.2 | 1 - 2 | 60 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |