Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501294.1 | XP_003501342.1 | 2 - 3 | 537 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |