Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506919.2 | XP_003506967.1 | 1 - 2 | 174 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |