Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508491.2 | XP_003508539.1 | 2 - 3 | 205 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |