Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il36b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515696.1 | XP_003515744.1 | 3 - 4 | 352 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |