Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641394.1 | XP_007639584.1 | 2 - 3 | 1142 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |