Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1ql2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499443.2 | XP_003499491.2 | 1 - 2 | 620 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |