Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647269.1 | XP_007645459.1 | 1 - 2 | 157 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |