Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508773.2 | XP_003508821.1 | 5 | 1123 | 116 | |
GenBank mRNA | JI889511.1 | - | 4 | 892 | 116 | |
JP055860.1 | - | 4 | 892 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |