Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APH1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677684.3 | XP_005677741.1 | 3 - 4 | 756 | 81 | |
GenBank mRNA | JO581146.1 | - | 3 - 4 | 686 | 81 | |
JT630049.1 | - | 490 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |