Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5MG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689515.3 | XP_005689572.1 | 2 - 3 | 470 | 104 | |
GenBank mRNA | JO580025.1 | - | 2 - 3 | 514 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |