Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLVAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682121.3 | XP_005682178.2 | 4 - 5 | 1338 | 96 | |
GenBank mRNA | JO579796.1 | - | 4 - 5 | 1455 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |