Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056690.1 | XP_017912179.1 | 4 - 5 | 1375 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |