Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10H15orf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685467.3 | XP_005685524.1 | 10 - 11 | 920 | 64 | |
XM_013966954.2 | XP_013822408.1 | 10 - 11 | 920 | 64 | ||
XM_013966957.2 | XP_013822411.1 | 11 - 12 | 959 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |