Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692931.2 | XP_005692988.2 | 6 - 7 | 2518 | 65 | |
XM_005692932.2 | XP_005692989.2 | 8 - 9 | 2518 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |