Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058798.1 | XP_017914287.1 | 5 - 6 | 888 | 77 | |
XM_018058807.1 | XP_017914296.1 | 5 - 6 | 888 | 77 | ||
XM_018058812.1 | XP_017914301.1 | 4 - 5 | 715 | 77 | ||
XM_018058819.1 | XP_017914308.1 | 4 - 5 | 628 | 77 | ||
XM_018058827.1 | XP_017914316.1 | 4 - 5 | 715 | 77 | ||
XM_018058837.1 | XP_017914326.1 | 5 - 6 | 888 | 77 | ||
XM_018058846.1 | XP_017914335.1 | 5 - 6 | 888 | 77 | ||
XM_018058847.1 | XP_017914336.1 | 4 - 5 | 715 | 77 | ||
XM_018058848.1 | XP_017914337.1 | 4 - 5 | 715 | 77 | ||
GenBank mRNA | KA318901.1 | - | 4 - 5 | 1203 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |