Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688516.3 | XP_005688573.2 | 2 - 3 | 868 | 92 | |
XM_005688517.3 | XP_005688574.2 | 2 - 3 | 906 | 92 | ||
XM_018057806.1 | XP_017913295.1 | 1 - 2 | 795 | 92 | ||
GenBank mRNA | JO438592.2 | - | 2 - 3 | 342 | 92 | |
JO603190.2 | - | 2 - 3 | 828 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |