Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRUNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677637.3 | XP_005677694.2 | 6 - 7 | 1901 | 86 | |
GenBank mRNA | JT669396.1 | - | 4 - 5 | 1484 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |