Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682787.2 | XP_005682844.1 | 2 - 3 | 732 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |