Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314273.1 | NP_001301202.1 | 2 - 3 | 1513 | 80 | |
XM_018058854.1 | XP_017914343.1 | 3 - 4 | 2018 | 80 | ||
XM_018058856.1 | XP_017914345.1 | 3 - 4 | 1737 | 80 | ||
XM_018058857.1 | XP_017914346.1 | 2 - 3 | 1569 | 80 | ||
GenBank mRNA | JX047313.1 | - | 2 - 3 | 1513 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |