Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696243.3 | XP_005696300.1 | 3 - 4 | 266 | 85 |
Gene Symbol: LOC102172949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048259.1 | XP_017903748.1 | 1 | 208 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |