Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNUPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695120.2 | XP_005695177.2 | 8 - 9 | 1160 | 72 | |
XM_018066121.1 | XP_017921610.1 | 8 - 9 | 862 | 72 | ||
XM_018066122.1 | XP_017921611.1 | 8 - 9 | 851 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |