Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDNEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693486.3 | XP_005693543.2 | 6 - 7 | 1310 | 101 | |
GenBank mRNA | JO411298.2 | - | 6 - 7 | 736 | 101 | |
JO498836.2 | - | 2 - 3 | 769 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |