Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDNEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693486.3 | XP_005693543.2 | 7 - 8 | 1392 | 64 | |
GenBank mRNA | JO411298.2 | - | 7 - 8 | 818 | 64 | |
JO498836.2 | - | 1 - 2 | 687 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |