Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CISH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695915.3 | XP_005695972.1 | 2 - 3 | 600 | 65 | |
GenBank mRNA | KA324707.1 | - | 1 - 2 | 1499 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |