Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285664.1 | NP_001272593.1 | 4 - 5 | 234 | 81 | |
GenBank mRNA | EU295697.1 | - | 4 - 5 | 234 | 81 | |
JO621471.2 | - | 4 - 5 | 305 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |