Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AURKAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690784.3 | XP_005690841.1 | 2 - 3 | 258 | 65 | |
XM_005690785.2 | XP_005690842.1 | 2 - 3 | 194 | 65 | ||
GenBank mRNA | JO353835.2 | - | 2 - 3 | 175 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |