Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53I3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686955.3 | XP_005687012.1 | 1 - 2 | 582 | 84 | |
XM_005686956.3 | XP_005687013.1 | 2 - 3 | 283 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |