Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287560.1 | NP_001274489.1 | 1 - 2 | 115 | 98 | |
GenBank mRNA | JQ417406.1 | - | 1 - 2 | 115 | 98 | |
JT700656.1 | - | 5 - 6 | 2160 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |