Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677179.3 | XP_005677236.1 | 3 - 4 | 340 | 63 | |
GenBank mRNA | JO452043.2 | - | 1 - 2 | 302 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |