Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057690.1 | XP_017913179.1 | 12 - 13 | 1613 | 61 | |
GenBank mRNA | KA351235.1 | - | 1 - 2 | 132 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |