Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057690.1 | XP_017913179.1 | 4 - 5 | 564 | 90 | |
GenBank mRNA | JO610655.2 | - | 1693 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |