Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044200.1 | XP_017899689.1 | 2 - 3 | 468 | 76 | |
GenBank mRNA | JO503505.2 | - | 1 - 2 | 796 | 76 | |
JO577190.1 | - | 2 - 3 | 508 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |