Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046977.1 | XP_017902466.1 | 2 - 3 | 1789 | 89 | |
GenBank mRNA | JO393717.2 | - | 1 - 2 | 288 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |