Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065196.1 | XP_017920685.1 | 2 - 3 | 397 | 61 | |
GenBank mRNA | JO611799.2 | - | 3 - 4 | 841 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |