Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056571.1 | XP_017912060.1 | 3 - 4 | 597 | 78 | |
XM_018056572.1 | XP_017912061.1 | 3 - 4 | 634 | 78 | ||
XM_018056573.1 | XP_017912062.1 | 3 - 4 | 550 | 78 | ||
XM_018056574.1 | XP_017912063.1 | 3 - 4 | 589 | 78 | ||
XM_018056575.1 | XP_017912064.1 | 3 - 4 | 405 | 78 | ||
XM_018056576.1 | XP_017912065.1 | 3 - 4 | 452 | 78 | ||
XM_018056577.1 | XP_017912066.1 | 3 - 4 | 459 | 78 | ||
XM_018056578.1 | XP_017912067.1 | 3 - 4 | 388 | 78 | ||
XM_018056579.1 | XP_017912068.1 | 3 - 4 | 599 | 78 | ||
GenBank mRNA | JO427973.1 | - | 3 - 4 | 633 | 78 | |
JO636128.2 | - | 3 - 4 | 388 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |