Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680682.3 | XP_005680739.1 | 3 - 4 | 1184 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |