Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047287.1 | XP_017902776.1 | 3 - 4 | 2410 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Capra hircus Probe spans exons |