Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056567.1 | XP_017912056.1 | 6 - 7 | 869 | 103 | |
GenBank mRNA | JO351699.2 | - | 4 - 5 | 374 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |