Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675932.3 | XP_005675989.1 | 1 - 2 | 833 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |