Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972144.2 | XP_013827598.1 | 3 - 4 | 407 | 90 | |
XM_013972145.2 | XP_013827599.1 | 2 - 3 | 340 | 90 | ||
XM_018063772.1 | XP_017919261.1 | 3 - 4 | 404 | 90 | ||
XM_018063773.1 | XP_017919262.1 | 3 - 4 | 407 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |