Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKIRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042069.1 | XP_017897558.1 | 4 - 5 | 708 | 94 | |
GenBank mRNA | KA346720.1 | - | 4 - 5 | 675 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |