Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22H3orf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695919.3 | XP_005695976.1 | 3 - 4 | 668 | 80 | |
XM_018038205.1 | XP_017893694.1 | 3 - 4 | 663 | 80 | ||
XM_018038206.1 | XP_017893695.1 | 3 - 4 | 487 | 80 | ||
GenBank mRNA | JO627990.2 | - | 2 - 3 | 1342 | 80 | |
JO627991.2 | - | 2 - 3 | 1342 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |