Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689577.3 | XP_005689634.3 | 3 - 4 | 2888 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |