Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691708.3 | XP_005691765.3 | 3 - 4 | 491 | 97 | |
GenBank mRNA | JT745148.1 | - | 3 - 4 | 370 | 97 | |
KA349629.1 | - | 3 - 4 | 487 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |